
Formic acid 98% ≥98%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03 g/mol
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8,II
:GHS05
Specification Test Results
- Assay:min. 98 %
- Acetic acid (CH3COOH):max. 0.4 %
- Ammonium (NH4):max. 0.005 %
- Color (APHA):max. 15
- Chloride (Cl):max. 0.001 %
- Iron (Fe):max. 0.001 %
- Heavy Metals (as Pb):max. 0.001 %
- Residue after Evaporation:max. 0.003 %
- Sulfate (SO4):max. 0.003 %
- Sulfite (SO3):passes test
Specification Test Results
Frequently Bought Together