Order Entry
Denmark
ContactUsLinkComponent
Formic acid ≥98.0%, ARISTAR® for trace analysis
Flammable
Corrosive
Toxic
Formic acid ≥98.0%, ARISTAR® for trace analysis
Catalog # 450122M
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Corrosive
Toxic
Formic acid ≥98.0%, ARISTAR® for trace analysis
Catalog # 450122M
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay (acidimetric):
    Min. 98.0 %
  • Colour:
    Max. 10 APHA
  • Cl (Chloride):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 0.5 ppm
  • Acetic acid:
    Max. 500 ppm
  • Ag (Silver):
    Max. 0.001 ppm
  • Al (Aluminium):
    Max. 0.005 ppm
  • Au (Gold):
    Max. 0.005 ppm
  • Ba (Barium):
    Max. 0.002 ppm
  • Be (Beryllium):
    Max. 0.001 ppm
  • Bi (Bismuth):
    Max. 0.002 ppm
  • Ca (Calcium):
    Max. 0.05 ppm
  • Cd (Cadmium):
    Max. 0.001 ppm
  • Co (Cobalt):
    Max. 0.001 ppm
  • Cr (Chromium):
    Max. 0.001 ppm
  • Cu (Copper):
    Max. 0.002 ppm
  • Fe (Iron):
    Max. 0.010 ppm
  • Ga (Gallium):
    Max. 0.005 ppm
  • Hg (Mercury):
    Max. 0.005 ppm
  • In (Indium):
    Max. 0.002 ppm
  • K (Potassium):
    Max. 0.02 ppm
  • Li (Lithium):
    Max. 0.001 ppm
  • Mg (Magnesium):
    Max. 0.01 ppm
  • Mn (Manganese):
    Max. 0.001 ppm
  • Mo (Molybdenum):
    Max. 0.001 ppm
  • Na (Sodium):
    Max. 0.02 ppm
  • Ni (Nickel):
    Max. 0.002 ppm
  • Pb (Lead):
    Max. 0.002 ppm
  • Sn (Tin):
    Max. 0.001 ppm
  • Sr (Strontium):
    Max. 0.001 ppm
  • Ti (Titanium):
    Max. 0.001 ppm
  • Tl (Thallium):
    Max. 0.001 ppm
  • V (Vanadium):
    Max. 0.001 ppm
  • Zn (Zinc):
    Max. 0.005 ppm
  • Non-volatile matter:
    Max. 2 ppm

Specification Test Results

About this item

 :
 :46.03 g/mol
Boiling Pt:100…101 °C (1013 hPa)
Melting Pt:8,2…8,4 °C
Density:1.22 g/cm³ (25 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8 (3),II
REACH:01-2119491174-37
 :GHS02
 :GHS05
 :GHS06
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297