Palmitic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:57-10-3
:
Formula:C₁₆H₃₂O₂
MDL Number:MFCD00002747
Merck Index:13,07064
Boiling Pt:352 °C (1013 hPa)
Melting Pt:61…63 °C
Density:0.853 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:256.43 g/mol
Storage Temperature:Ambient
EINECS:200-312-9
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:61 - 63 °C
- Molar Mass:256.42 g/mol
- Bulk Density:415 kg/m3
- Boiling Point:271.5 °C (133 hPa)
- Flash Point:206 °C
- Density:0.852 g/cm3 (20 °C)
Specification Test Results
Related Information
Frequently Bought Together