Order Entry
Denmark
ContactUsLinkComponent
Acetic acid glacial ≥98%, TECHNICAL
Flammable
Corrosive
Acetic acid glacial ≥98%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Corrosive
Acetic acid glacial ≥98%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

Frequently Bought Together