Supelco® Organic Reference Standard, Isopentyl acetate
Supplier: Merck
Certificates
About this item
- GHS signal: GHS02
Caution: P210
Specifications
- CAS No.:123-92-2
- Storage conditions:2...30 °C
- Hazard class description:H226
- Shipping conditions:STD
- MDL No.:MFCD00008946
- M-Clarity quality level:MQ200
- Synonym(s):Acetic acid 3-methylbutyl ester; Isoamyl acetate
- Melting point:–78 °C (lit.)
- Formula:CH₃COOCH₂CH₂CH(CH₃)₂
- Molecular weight:130,18
- Density:0,876 g/ml at 25 °C (lit.)
- Flash point:33 °C - Closed cup
- Boiling point:142 °C/756 mmHg (lit.)
Specifications
Frequently Bought Together