Order Entry
Denmark
ContactUsLinkComponent
Sputum containers, Nunc™
  391-7798
   Low Manufacturing Impact
undefined
Sputum containers, Nunc™
  391-7798
   Low Manufacturing Impact
  • Ext.Ø:
    80 mm
  • Packed:
    Case of 150
  • Type:
    Non sterile
  • Capacity:
    24 ml
  • Height:
    28,5 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
Polystyrol.
  • Low profile container can be shipped by normal mail
  • Large opening facilitates secure sample placement
  • Lowered centre of lid facilitates scraping off samples when the lid is removed
  • Ribs on lid and container for easy grip
For sampling and transport of diagnostic specimens.
 : Containers comply with 95 kPa test described in UN packaging instructions 602/650 for transport of infectious substances and diagnostic specimens.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904.