Bottles, wide neck, square with thread
: MAUSER
HDPE, translucent or HDPE, white.
- Good chemical resistance
- Resistant to temperatures from −40 to +110 °C
- Tamper-evident closures must be ordered separately (see below for available options)
Ideal for collecting, storing, and shipping liquids, pastes, and granulates. Square shape saves space, recessed labelling space. Wide neck makes filling and cleaning easy.
Frequently Bought Together