Ordreindtastning
Denmark
ContactUsLinkComponent
2-Nonanon, Supelco®
2-Nonanon, Supelco®
Catalog # 63969-1ML
Supplier:  Merck
CAS Number:  
undefined
2-Nonanon, Supelco®
Catalog # 63969-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :142.24 g/mol
Boiling Pt:192 °C (1013 hPa)
Melting Pt:–15 °C
Density:0,82 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Stue temperatur
EINECS:212-480-0
CAS Number:821-55-6
Formula:C₉H₁₈O
MDL Number:MFCD00009553