2-Nonanone
Catalog # EHERC15623220
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:142.24 g/mol
Boiling Pt:192 °C (1013 hPa)
Melting Pt:–15 °C
Density:0.82 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-480-0
CAS Number:821-55-6
Formula:C₉H₁₈O
MDL Number:MFCD00009553