N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Catalog # HYPETS-48913
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:199.25 g/mol
Boiling Pt:130 °C (1013 hPa)
Density:1,07 g/cm³ (20 °C)
EINECS:246-331-6
:GHS02
:GHS07
CAS Number:24589-78-4
Formula:C₆H₁₂F₃NOSi
MDL Number:MFCD00000411
Boiling Pt:130 °C (1013 hPa)