Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-(2-Ethoxyethoxy)ethyl acetate ≥99%

2-(2-Ethoxyethoxy)ethyl acetate ≥99%

2-(2-Ethoxyethoxy)ethyl acetate ≥99%
Certificates
2-(2-Ethoxyethoxy)ethyl acetate ≥99%
2-(2-Ethoxyethoxy)ethyl acetate ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 112-15-2 Molecular Formula: CH3CO2(CH2CH2O)2C2H5 Formula Weight: 176.21 Flash Point: 95°(203°F) Refractive Index: 1.4210 Melting Point: -25° Boiling Point: 218-219° Beilstein Reference: 1764643 Merck Reference: 14,1800 MDL No.: MFCD00041928

Synonyms:  Diethylene glycol monoethyl ether acetate
  • CAS Number:112-15-2
  • Formula:C₈H₁₆O₄
  • MDL Number:MFCD00041928
  • Molecular Weight:176.21 g/mol
  • Merck Index:14,1800
  • Boiling Pt:218…219 °C
  • Melting Pt:–25 °C
  • Density:1.012
  • Flash Pt:95 °C (203 °F)
  • Storage Temperature:Ambient
  • EINECS:203-940-1
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.