Order Entry
Denmark
Orders LinkContactUsLinkComponent
Apigenin, Millipore®

Apigenin, Millipore®

Glass bottle for solids, 5 mg
Catalog # 178278-5
Apigenin, Millipore®
Catalog # 178278-5
CAS Number:  520-36-5
Apigenin, Millipore®

Glass bottle for solids, 5 mg

CAS Number:  520-36-5
Price
353,00 kr
Log in for availability
1 * 5 mg
Product Details & Documents
Synonyms:  4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
  • CAS Number:520-36-5
  • Formula:C₁₅H₁₀O₅
  • MDL Number:MFCD00006831
  • Molecular Weight:270.24 g/mol
  • EINECS:208-292-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg

Specifications