Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 3-Chloropropene 2000 µg/ml in methanol
Organic reference standard, 3-Chloropropene 2000 µg/ml in methanol
Supplier:  Restek
Certificates
Organic reference standard, 3-Chloropropene 2000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    107-05-1
  • Formula:
    C₃H₅Cl
  • Description:
    3-Chloropropene

Specifications

Frequently Bought Together