Order Entry
Denmark
Orders LinkContactUsLinkComponent
Acetonitrile

Acetonitrile

Flammable
Irritant
   Sustainable Options Available
Marketsource
Supplier:   Roth Carl
Acetonitrile
Supplier:  Roth Carl
   Sustainable Options Available
Marketsource
Certificates
Acetonitrile

2 Options Available Below

Product Details & Documents
Synonyms:  Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
  • CAS Number:75-05-8
  • Formula:H₃CCN
  • MDL Number:MFCD00001878
  • Molecular Weight:41.05 g/mol
  • Merck Index:13,00071
  • Boiling Pt:81.6 °C (1013 hPa)
  • Melting Pt:–45.7 °C
  • Density:0.782 g/cm³ (20 °C)
  • Flash Pt:2 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-835-2
  • UN:1648
  • ADR:3,II
  • REACH:01-2119471307-38
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ROTH7330.1
  • ROTH7330.2
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

    Specifications:
      Safer Human and Environmental Health
    Marketsource
    More
    Specifications
    Cat. No.
    ROTH7330.1
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
    Marketsource
  • Plastic bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
      Safer Human and Environmental Health
    Marketsource
    More
    Specifications
    Cat. No.
    ROTH7330.2
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
    Marketsource