Order Entry
Denmark
Orders LinkContactUsLinkComponent
Baicalein ≥97%
Baicalein ≥97%
Baicalein ≥97%
Baicalein ≥97%
Baicalein ≥97%
Purity: 97% CAS No.: 491-67-8 Molecular Formula: C15H10O5 Formula Weight: 270.24 Melting Point: 268-270° Beilstein Reference: 272683 Merck Reference: 14,942 MDL No.: MFCD00017459
 :  5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
  •  :491-67-8
  •  :C₁₅H₁₀O₅
  •  :MFCD00017459
  •  :270.24 g/mol
  •  :14,942
  •  :268…270 °C
  •  :Ambient
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    100 mg
    1 * 100 mg
    1 * 100 mg - 526,00 kr
    526,00 kr
    1 * 100 mg
       526,00 kr
  • Glass bottle for solids
    500 mg
    1 * 500 mg
    1 * 500 mg - 1.690,00 kr
    1.690,00 kr
    1 * 500 mg
       1.690,00 kr