Order Entry
Denmark
Orders LinkContactUsLinkComponent
Baicalein

Baicalein

Supplier:   Cayman Chemical
Baicalein
Supplier:  Cayman Chemical
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
  • CAS Number:491-67-8
  • Formula:C₁₅H₁₀O₅
  • MDL Number:MFCD00017459
  • Molecular Weight:270.24 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 224,00 kr
    224,00 kr
    1 * 50 mg
       224,00 kr

  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 395,00 kr
    395,00 kr
    1 * 100 mg
       395,00 kr

  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 1.570,00 kr
    1.570,00 kr
    1 * 500 mg
       1.570,00 kr

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 2.710,00 kr
    2.710,00 kr
    1 * 1 g
       2.710,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.