2-Ethylhexane-1,3-diol (mixture of isomers) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:94-96-2
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00004578
:146.23 g/mol
Boiling Pt:244 °C (1013 hPa)
Melting Pt:–40 °C
Density:0.9422 g/cm³
Flash Pt:136 °C
EINECS:202-377-9
:GHS05
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4495 to 1.4525 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥98.5 %
- Color scale:≤15 APHA
- Acidity:≤0.05 mg KOH/g
Specification Test Results
Frequently Bought Together