Order Entry
Denmark
ContactUsLinkComponent
1-Pentanol
1-Pentanol
Catalog # ROTH8949.1
Supplier:  Roth Carl
CAS Number:  
undefined
1-Pentanol
Catalog # ROTH8949.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

About this item

 :
 :88.15 g/mol
Merck Index:13,07195
Boiling Pt:137.9 °C (1013 hPa)
Melting Pt:–79 °C
Density:0.81 g/cm³ (20 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:200-752-1
UN:1105
ADR:3,III
 :GHS02
 :GHS07
CAS Number:71-41-0
Formula:H₃C(CH₂)₃CH₂OH
MDL Number:MFCD00002977