2-Chlorophenol-[ring-D₄] (99% D)
Glass bottle for solids, 0,1 g
Catalog # CAMBDLM-1638-0.1
Supplier: CAMBRIDGE ISOTOPE LABS
2-Chlorophenol-[ring-D₄] (99% D)
Catalog # CAMBDLM-1638-0.1
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number: 93951-73-6
Jump to:
Product Details & Documents
Synonyms: 2-Chlorophenol-[ring-²H₄], 2-Chlorophenol (Ring-D4), 1-Chloro-2-hydroxybenzene-D4
- CAS Number:93951-73-6
- Formula:C₆HD₄ClO
- Molecular Weight:132.53
- Storage Temperature:Ambient
- UN:2021
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



