Amantadine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride
CAS Number:665-66-7
:
Formula:C₁₀H₁₈ClN
MDL Number:MFCD00074723
Molecular Weight:187.71 g/mol
Melting Pt:360
:M-Clarity™ quality level = MQ200
EINECS:211-560-2
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:360 °C
- Molar Mass:187.71 g/mol
Specification Test Results
Frequently Bought Together