Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, prednisolones, Ph. Eur., RS
Organic reference standards, prednisolones, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, prednisolones, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    52-21-1
  • Formula:
    C₂₃H₃₀O₆
  • Description:
    Prednisolone acetate for peak identification

Specifications

Frequently Bought Together