Order Entry
Denmark
ContactUsLinkComponent
1,2-Dichloro-3-nitrobenzene
  EHERC12432300
 :  
1,2-Dichloro-3-nitrobenzene
  EHERC12432300
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029
 :
 :192 g/mol
 :257 °C (1013 hPa)
 :61 °C
 :1.721 g/cm³ (14 °C)
 :Ambient
 :221-717-7
 :1578
 :6.1,II
 :3209-22-1
 :C₆H₃Cl₂NO₂
 :MFCD00007062