Order Entry
Denmark
ContactUsLinkComponent
Triphosgene
Triphosgene
Catalog # MOLEM19441560
Supplier:  Molekula
CAS Number:  
Triphosgene
Catalog # MOLEM19441560
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :296.75 g/mol
Boiling Pt:203 °C (1013 hPa)
Melting Pt:77…81 °C
Density:1.629 g/cm³ (80 °C)
EINECS:250-986-3
UN:2928
ADR:6.1,II
CAS Number:32315-10-9
Formula:C₃Cl₆O₃
MDL Number:MFCD00062848