

Triphosgene 99%
Leverandør: Thermo Fisher Scientific
Certifikater
Om denne vare
CAS nummer:32315-10-9
:
Formel:C₃Cl₆O₃
MDL nummer:MFCD00062848
:296.75 g/mol
Kogende Pt:203…206 °C (760 mmHg)
Smeltende Pt:77…81 °C
Tæthed:1.78 g/cm³
Opbevaringstemperatur:Refrigerator
EINECS:250-986-3
FN:2928
ADR:6.1,II
:GHS05
:GHS06
Specifikationstestresultater
- Appearance:White to off-white Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:77 to 81 °C
- Assay:≥98.5 %
Specifikationstestresultater
Frequently Bought Together