1,4-Dithioerythritol
Marketsource
Supplier: Serva
Jump to:
About this item
Synonyms: Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
- CAS Number:6892-68-8
- Formula:C₄H₁₀O₂S₂
- MDL Number:MFCD00063750
- Molecular Weight:154.25 g/mol
- Boiling Pt:286 °C (1013 hPa)
- Melting Pt:82…83 °C
- Storage Temperature:Ambient
- EINECS:229-998-8
3 Options Available Below
Product Details & Documents
Synonyms: Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
- CAS Number:6892-68-8
- Formula:C₄H₁₀O₂S₂
- MDL Number:MFCD00063750
- Molecular Weight:154.25 g/mol
- Boiling Pt:286 °C (1013 hPa)
- Melting Pt:82…83 °C
- Storage Temperature:Ambient
- EINECS:229-998-8
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



