Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for Corning® E-Cube™
Screw cap, 45 mm, with two stainless steel tubing ports, for E-Cube™ Culture System, PP
  734-4184
 :   Corning
Accessories for Corning® E-Cube™
  734-4184
 :  Corning
Accessories for Corning® E-Cube™
Accessories for Corning® E-Cube™
Screw cap, 45 mm, with two stainless steel tubing ports, for E-Cube™ Culture System, PP
1.890,00 kr
1 * 1 Items
  • Description:
    Screw cap, 45 mm, with two stainless steel tubing ports, for E-Cube™ Culture System
  • Material:
    PP