
1,5-Diaminopentane ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:462-94-2
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00008239
:102.18 g/mol
Merck Index:14,01609
Boiling Pt:180°C
Density:0,9174 g/cm³ (0 °C)
Flash Pt:63°C
Storage Temperature:Ambient
EINECS:207-329-0
UN:2735
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together