

Zinc(II) nitrate hexahydrate ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10196-18-6
:
Formula:ZnN₂O₆·6H₂O
MDL Number:MFCD00149889
:297.49 g/mol
Merck Index:14,10144
Boiling Pt:∼105 °C (1013 hPa)
Melting Pt:∼36 °C
Density:2.07
Storage Temperature:Ambient
EINECS:231-943-8
UN:1514
ADR:5.1,II
:GHS03
:GHS07
Frequently Bought Together