Sodium thiosulphate pentahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:10102-17-7
:
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186
:248.19 g/mol
Merck Index:12,08844
Melting Pt:48 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-867-5
Frequently Bought Together