Order Entry
Denmark
ContactUsLinkComponent
Citric acid monohydrate 99.5-100.5%, powder Ph. Eur.
Irritant
Citric acid monohydrate 99.5-100.5%, powder Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Citric acid monohydrate 99.5-100.5%, powder Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on anhydrous):
    99.5 - 100.5 %
  • Appearance:
    Conforms (see CoA/CoS)
  • Identification B:
    Passes test
  • Appearance of solution:
    Passes test
  • Readily carbonisable substances:
    Passes test
  • Oxalic acid (calculated on anhydrous):
    Max. 360 ppm
  • SO4 (Sulphate):
    Max. 150 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Water:
    7.5 - 9.0 %
  • Sulphated ash:
    Max. 0.1 %
  • Residual solvents:
    Unlikely by manuf.process

Specification Test Results

Frequently Bought Together