Order Entry
Denmark
ContactUsLinkComponent
Citric acid monohydrate 99.7-100.5%, AnalaR NORMAPUR® ACS, Reag. USP, Reag. Ph. Eur. analytical reagent
Irritant
Citric acid monohydrate 99.7-100.5%, AnalaR NORMAPUR® ACS, Reag. USP, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Citric acid monohydrate 99.7-100.5%, AnalaR NORMAPUR® ACS, Reag. USP, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    99.7 - 100.5 %
  • Identification (IR):
    Passes test
  • Readily carbonisable substances:
    Passes test
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.02 %
  • Insolubility in water:
    Max. 50 ppm
  • Oxalic acid (calculated on anhydrous):
    Max. 360 ppm
  • Tartaric acid:
    Max. 0.2 %
  • Water:
    7.5 - 9.0 %
  • C2O4 (as C2H2O4):
    Max. 0.04 %
  • Cl (Chloride):
    Max. 5 ppm
  • PO4 (Phosphate):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • As (Arsenic):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 20 ppm
  • Cu (Copper):
    Max. 5 ppm
  • Fe (Iron):
    Max. 3 ppm
  • Mg (Magnesium):
    Max. 5 ppm
  • Ni (Nickel):
    Max. 1 ppm
  • Pb (Lead):
    Max. 2 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test
  • Conforms to Reag. USP:
    Passes test

Specification Test Results

Frequently Bought Together