Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, (R)-(+)-Limonene
Supelco® Organic Reference Standard, (R)-(+)-Limonene
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, (R)-(+)-Limonene
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    5989-27-5
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H226 - H304 - H315 - H317 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00062991
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (+)-p-Mentha-1,8-diene; (+)-Carvene; (R)-4-Isopropenyl-1-methyl-1-cyclohexene
  • Molecular weight:
    136,23
  • Density:
    0,842 g/ml at 20 °C (lit.)
  • Flash point:
    51 °C - closed cup
  • Boiling point:
    176...177 °C (lit.)

Specifications

Related Information

Frequently Bought Together