Sodium hydroxide 2 mol/l (2 N) in aqueous solution 2 M NaOH (2N) volumetric solution, Fluka™
Plastic bottle, 1 L
Supplier: Honeywell Chemicals
Sodium hydroxide 2 mol/l (2 N) in aqueous solution 2 M NaOH (2N) volumetric solution, Fluka™
Catalog # 35254-1L
Supplier: Honeywell Chemicals
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Honeywell's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)Buffers available as concentrates or ready-to-useReagents for complexometry:Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
UNSPSC Code: 12352316
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:105…140 °C (1.013 hPa)
- Melting Pt:–12 – –10 °C.
- Density:1.08 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Documents
Specifications
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specifications
- Pack type:Plastic bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



