Order Entry
Denmark
ContactUsLinkComponent
Perchloric acid standardised solution 0.1 N in acetic acid
Corrosive
Flammable
Irritant
Perchloric acid standardised solution 0.1 N in acetic acid
Catalog # 35608.K2
CAS Number:  
undefined
Corrosive
Flammable
Irritant
Perchloric acid standardised solution 0.1 N in acetic acid
Catalog # 35608.K2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

About this item

CAS Number:7601-90-3
 :
Formula:HClO₄
MDL Number:MFCD00011325
 :100.46 g/mol
Storage Temperature:Ambient
EINECS:231-512-4
UN:2920
ADR:8,II
 :GHS05
 :GHS02
 :GHS07