Order Entry
Denmark
ContactUsLinkComponent
Perchloric acid 0.01 mol/l (0.01 N) in anhydrous acetic acid
 
 
Perchloric acid 0.01 mol/l (0.01 N) in anhydrous acetic acid
  87116.290
 :  
undefined
 
 
Perchloric acid 0.01 mol/l (0.01 N) in anhydrous acetic acid
  87116.290
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

 :
 :100.46 g/mol
 :Ambient
 :231-512-4
 :2789
 :8,II
 :GHS02
 :GHS05
 :7601-90-3
 :HClO₄
 :MFCD00011325