Order Entry
Denmark
ContactUsLinkComponent
Sodium chloride 1.0 M NaCl (1.0N) volumetric solution, Fluka™
Sodium chloride 1.0 M NaCl (1.0N) volumetric solution, Fluka™
Catalog # 71394-1L
CAS Number:  
Sodium chloride 1.0 M NaCl (1.0N) volumetric solution, Fluka™
Catalog # 71394-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Density (D 20/4):
    1.030 - 1.050
  • Factor:
    0.995 - 1.005

Specification Test Results

About this item

 :
 :58.44 g/mol
Density:1.00 g/cm³
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477