Sodium hydroxide 1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09956.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- Seq:0501
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.030 g/cm3 (20 °C)
- pH:14 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
Synonyms: Caustic soda
:
:40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
EINECS:215-185-5
UN:1824
:GHS05
Storage Temperature:Ambient
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548
Melting Pt:323
Density:1.327 g/cm³ (20 °C)
ADR:8
Merck Index:14,08627
:M-Clarity™ quality level = MQ100