Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

(±)-2-Hexanol 99%

Flammable
Irritant
(±)-2-Hexanol 99%
Certificates
(±)-2-Hexanol 99%

3 Options Available Below

Product Details & Documents
Synonyms:  (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane
  • CAS Number:626-93-7
  • Formula:C₆H₁₄O
  • MDL Number:MFCD00004585
  • Molecular Weight:102.18 g/mol
  • Boiling Pt:139 °C (760 mmHg)
  • Melting Pt:–23 °C
  • Density:0.81 g/cm³
  • Flash Pt:41 °C
  • Storage Temperature:Ambient
  • EINECS:210-971-4
  • UN:2282
  • ADR:3,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4125 to 1.4155 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information