Osmiumtetraoxid 4% (w/w) in water
:20816-12-0
:
:O₄Os
:MFCD00011150
:254.23 g/mol
:1,04 g/cm³
:Køleskab
:244-058-7
:3287
:6.1,II
:GHS06
:GHS05
:GHS08
- Appearance:Clear yellow Solution
- Strength:3.92 to 4.08 %
Frequently Bought Together