Order Entry
Denmark
ContactUsLinkComponent
 
Hydrogen peroxide 31% unstabilised, VLSI Selectipur® for the electronics industry
Catalog # 50489142.
CAS Number:  
undefined
Hydrogen peroxide 31% unstabilised, VLSI Selectipur® for the electronics industry
Catalog # 50489142.
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
Melting Pt:? 0 °C
EINECS:0
ADR:5.1 (8),II
 :34.01 g/mol
UN:2014
 :GHS03
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333