Order Entry
Denmark
Orders LinkContactUsLinkComponent
Carboys, narrow neck, PC, Biotainer®, Nalgene®
Carboys, narrow neck, PC, Biotainer®, Nalgene®
   Sustainable Options Available
Carboys, narrow neck, PC, Biotainer®, Nalgene®
   Sustainable Options Available
Certificates
Carboys, narrow neck, PC, Biotainer®, Nalgene®
Carboys, narrow neck, PC, Biotainer®, Nalgene®
6 Options Available Below
Product Details & Documents
PC, clear, with white screw cap.
  • Silicone-lined polypropylene screw cap
  • Good chemical resistance
  • Good temperature resistance
Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations. Use sterile, ready-to-use Biotainer™ Carboys, designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.
Specifications for Product Family
Specifications
  • Catalog No:
  • 391-7103
  • 391-7102
  • 391-7104
  • 391-7100
  • 391-7101
  • NALG3405-99
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Type
    Ext.Ø
    Neck I-Ø
    Height
    Thread
    Availability
    Price
  • 10 l
    with handle, individually wrapped
    250 mm
    37 mm
    337 mm
    48 mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 1.430,00 kr
    1.430,00 kr
    1 * 2 Items
       1.430,00 kr
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-7103
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 10 l
    with handle, bulk packed
    250 mm
    37 mm
    337 mm
    48 mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 3.200,00 kr
    3.200,00 kr
    1 * 2 Items
       3.200,00 kr
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-7102
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 20 l
    with handle, individually wrapped
    255 mm
    37 mm
    494 mm
    48 mm
    Log in for availability
    Log in for availability
    1 * 3 Items
    1 * 3 Items - 4.060,00 kr
    4.060,00 kr
    1 * 3 Items
       4.060,00 kr
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-7104
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 5 l
    with handle, individually wrapped
    166 mm
    38 mm
    290 mm
    48 mm
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 3.570,00 kr
    3.570,00 kr
    1 * 6 Items
       3.570,00 kr
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-7100
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 5 l
    without handle, individulla wrapped
    166 mm
    38 mm
    290 mm
    48 mm
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 4.310,00 kr
    4.310,00 kr
    1 * 6 Items
       4.310,00 kr
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-7101
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 5 l
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items
    Specifications:
    More
    Specifications
    Cat. No.
    NALG3405-99
    Environmentally Preferable
    Environmental certification