

Hydrochloric acid 32 - 35%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:42 °C (1.013 hPa)
Melting Pt:–35 °C
Density:1.190 g/cm³ (20 °C)
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Assay:32.0 - 35.0 %
- Density (D 20/4):1.145 - 1.180
- Silver (Ag):Max. 10 ppt
- Aluminium (Al):Max. 50 ppt
- Arsenic (As):Max. 100 ppt
- Gold (Au):Max. 20 ppt
- Barium (Ba):Max. 10 ppt
- Beryllium (Be):Max. 10 ppt
- Bismuth (Bi):Max. 10 ppt
- Calcium (Ca):Max. 200 ppt
- Cadmium (Cd):Max. 10 ppt
- Cerium (Ce):Max. 10 ppt
- Cobalt (Co):Max. 10 ppt
- Chromium (Cr):Max. 100 ppt
- Cesium (Cs):Max. 10 ppt
- Copper (Cu):Max. 100 ppt
- Dysprosium (Dy):Max. 10 ppt
- Iron (Fe):Max. 200 ppt
- Erbium (Er):Max. 10 ppt
- Europium (Eu):Max. 10 ppt
- Gallium (Ga):Max. 10 ppt
- Gadolinium (Gd):Max. 10 ppt
- Hafnium (Hf):Max. 10 ppt
- Mercury (Hg):Max. 500 ppt
- Holmium (Ho):Max. 10 ppt
- Indium (In):Max. 10 ppt
- Potassium (K):Max. 100 ppt
- Lanthanum (La):Max. 10 ppt
- Lithium (Li):Max. 10 ppt
- Lutetium (Lu):Max. 10 ppt
- Magnesium (Mg):Max. 50 ppt
- Manganese (Mn):Max. 10 ppt
- Molybdenum (Mo):Max. 50 ppt
- Sodium (Na):Max. 100 ppt
- Niobium (nb):Max. 10 ppt
- Neodymium (nd):Max. 10 ppt
- Nickel (Ni):Max. 50 ppt
- Lead (Pb):Max. 10 ppt
- Palladium (Pd):Max. 10 ppt
- Praseodymium (Pr):Max. 10 ppt
- Platinum (Pt):Max. 10 ppt
- Rubidium (Rb):Max. 10 ppt
- Rhenium (Re):Max. 10 ppt
- Rhodium (Rh):Max. 10 ppt
- Ruthenium (Ru):Max. 10 ppt
- Antimony (Sb):Max. 100 ppt
- Scandium (Sc):Max. 20 ppt
- Selenium (Se):Max. 1000 ppt
- Samarium (Sm):Max. 10 ppt
- Tin (Sn):Max. 200 ppt
- Strontium (Sr):Max. 10 ppt
- Terbium (Tb):Max. 10 ppt
- Tellurium (Te):Max. 10 ppt
- Thorium (Th):Max. 10 ppt
- Titanium (Ti):Max. 50 ppt
- Thallium (Tl):Max. 10 ppt
- Thulium (Tm):Max. 10 ppt
- Uran (U):Max. 10 ppt
- Vanadium (V):Max. 10 ppt
- Tungsten (W):Max. 10 ppt
- Yttrium (Y):Max. 10 ppt
- Ytterbium (Yb):Max. 10 ppt
- Zinc (Zn):Max. 100 ppt
- Zirconium (Zr):Max. 10 ppt
- Vapor Pressure:3.23 psi ( 21.1 °C)
Specification Test Results
Frequently Bought Together