


Naphthalene-[D₈] (98% D) for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1146-65-2
:
Formula:C₁₀D₈
MDL Number:MFCD00001743
:136.11 g/mol
Melting Pt:81…83 °C
Flash Pt:78 °C
EINECS:214-552-7
UN:1334
ADR:4.1,III
:GHS07
:GHS02
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals
- D-Enrichment:≥98.0 %
- NMR (19F-NMR):≥96.0 %
Specification Test Results
Frequently Bought Together