Order Entry
Denmark
ContactUsLinkComponent
Chlorobenzene-[D₅] 98.5+ atom % D for NMR spectroscopy
Irritant
Environment
Flammable
Chlorobenzene-[D₅] 98.5+ atom % D for NMR spectroscopy
Certificates
undefined
Irritant
Environment
Flammable
Chlorobenzene-[D₅] 98.5+ atom % D for NMR spectroscopy
Certificates
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 %
  • D-Enrichment:
    ≥98.50 %
  • HPLC:
    ≥97.5 %