



n-Hexane-[D₁₄] (98% D)
Catalog # CAMBDLM-139-5
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:100.07 g/mol
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.96 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:21666-38-6
Formula:D₃C(CD₂)₄CD₃
MDL Number:MFCD00037562