Order Entry
Denmark
ContactUsLinkComponent
n-Hexane-[D₁₄] (99% D) for NMR spectroscopy
Flammable
Health
Irritant
Environment
n-Hexane-[D₁₄] (99% D) for NMR spectroscopy
Catalog # 8.15043.0005
Supplier:  Merck
CAS Number:  
Flammable
Health
Irritant
Environment
n-Hexane-[D₁₄] (99% D) for NMR spectroscopy
Catalog # 8.15043.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Solubility:
    0.0095 g/l (20 °C)
  • Melting Point:
    -94.3 °C
  • Molar Mass:
    100.26 g/mol
  • Boiling Point:
    69 °C (1013 hPa)
  • Vapor Pressure:
    160 hPa (20 °C)
  • Flash Point:
    -22 °C
  • Explosion Limit:
    1.0 - 8.1 %(V)
  • Density:
    0.767 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    240 °C DIN 51794
  • Viscosity kinematic:
    0.50 mm2/s (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

About this item

 :
 :100.07 g/mol
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.96 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1208
ADR:3,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:21666-38-6
Formula:D₃C(CD₂)₄CD₃
MDL Number:MFCD00037562