

Iodomethane[D₃] (99% D) for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:865-50-9
:
Formula:D₃CI
MDL Number:MFCD00001074
:144.92 g/mol
Boiling Pt:42 °C (1013 hPa)
Density:2.329 g/cm³
Storage Temperature:Refrigerator
EINECS:212-744-5
UN:2644
ADR:6.1,I
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear colorless Liquid (May darken on storage)
- Water:≤0.05 % (Coulometric)
- D-Enrichment:≥99.0 %
- HPLC:≥98.0 %
Specification Test Results
Frequently Bought Together