Chloroform-[D₁] 100.0 atom % D for NMR spectroscopy
:865-49-6
:
:CDCl₃
:MFCD00000827
:120.38 g/mol
:60.8 °C (1013 hPa)
:–64 °C
:1.5 g/cm³ (20 °C)
:Refrigerator
:212-742-4
:1888
:6.1,III
:GHS06
:GHS08
- Appearance:Clear colorless Liquid
- Water:≤0.01 % (Coulometric)
- D-Enrichment:≥99.95 %
- GC:≥96.0 %
Frequently Bought Together