Order Entry
Denmark
ContactUsLinkComponent
Dichloromethane-[D₂] (99.8% D) for NMR spectroscopy, Sigma-Aldrich®
Health
Irritant
Dichloromethane-[D₂] (99.8% D) for NMR spectroscopy, Sigma-Aldrich®
Catalog # 1.13720.0009
Supplier:  Merck
CAS Number:  
Health
Irritant
Dichloromethane-[D₂] (99.8% D) for NMR spectroscopy, Sigma-Aldrich®
Catalog # 1.13720.0009
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    7,5 mL
  • Seq:
    2260

Specifications

About this item

 :
 :86.92 g/mol
Boiling Pt:40 °C (1013 hPa)
Melting Pt:–97 °C
Density:1.36 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-776-0
UN:1593
ADR:6.1,
 :M-Clarity™ quality level = MQ200
 :GHS08
 :GHS07
CAS Number:1665-00-5
Formula:CD₂Cl₂
MDL Number:MFCD00000882