Order Entry
Denmark
ContactUsLinkComponent
 
 
 
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
  ACRO320720075
 :  
undefined
 
 
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
  ACRO320720075
 :  

 

  • Pack type:
    Ampoule
  • Pk:
    7,5 mL
  • Seq:
    2065
  • Pk Info:
    10 x 0.75 ml

 

 

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %

 

 

 :
 :86.92 g/mol
 :40 °C (1013 hPa)
 :–97 °C
 :1.36 g/cm³ (20 °C)
 :Ambient
 :216-776-0
 :1593
 :6.1,III
 :GHS07
 :GHS08
 :1665-00-5
 :CD₂Cl₂
 :MFCD00000882